Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-1-(4-Chlorophenyl)ethyl isocyanate, tech. 90%

Chemical Identifiers

Chemical Name or Material (S)-(-)-1-(4-Chlorophenyl)ethyl isocyanate
CAS 745783-72-6
Purity Grade Technical Grade
Molecular Formula C9H8ClNO
Formula Weight 181.62
MDL Number MFCD05664072
UN Number UN2206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2022703

 
alfa aesar™
L2022703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.62
Chemical Name or Material (S)-(-)-1-(4-Chlorophenyl)ethyl isocyanate
Molecular Formula C9H8ClNO
MDL Number MFCD05664072
CAS 745783-72-6
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Purity Grade Technical Grade
Assay 90%
UN Number UN2206
Health Hazard 1 Danger
Quantity 1g
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C