Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-1-(4-Bromophenyl)ethylamine, ChiPros, 99%, ee 98%

Chemical Identifiers

Chemical Name or Material (S)-(-)-1-(4-Bromophenyl)ethylamine
CAS 27298-97-1
Molecular Formula C8H10BrN
Formula Weight 200.08
MDL Number MFCD00066026
UN Number 2735
Beilstein 5729988
Synonym (S)-(-)-p-Bromo-alpha-methylbenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2008003

 
alfa aesar™
L2008003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2008006

 
alfa aesar™
L2008006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2008014

 
alfa aesar™
L2008014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.08
Chemical Name or Material (S)-(-)-1-(4-Bromophenyl)ethylamine
MDL Number MFCD00066026
Assay 99%
Molecular Formula C8H10BrN
CAS 27298-97-1
UN Number UN2735
Beilstein 5729988
Synonym (S)-(-)-p-Bromo-alpha-methylbenzylamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Assay Percent Notes ee 98%
Boiling Point 140° to 145°C (30mmHg)
Melting Point −25°C (neat)
Quantity 1g
Flash Point >110°C (230°F)
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Applications
(S)-(-)-1-(4-Bromophenyl)ethylamine is utilized as an important raw material in organic synthesis. It also serves as an intermediate in organic synthesis and pharmaceuticals and agrochemicals.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place.

Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.