Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-1-(3-Chlorophenyl)ethylamine, ChiPros™, 98%, ee 98+%

Chemical Identifiers

Chemical Name or Material (S)-1-(3-Chlorophenyl)ethylamine
CAS 68297-62-1
Molecular Formula C8H10ClN
Formula Weight 155.63
MDL Number MFCD06761823
UN Number UN2735
Synonym (S)-3-Chloro-alpha-methylbenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2763703

  SDS
alfa aesar™
H2763703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2763706

  SDS
alfa aesar™
H2763706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 155.63
Chemical Name or Material (S)-1-(3-Chlorophenyl)ethylamine
Assay 98%
Assay Percent Notes ee ≥98%
Molecular Formula C8H10ClN
MDL Number MFCD06761823
CAS 68297-62-1
UN Number UN2735
Synonym (S)-3-Chloro-alpha-methylbenzylamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 112°C (13mmHg)
Quantity 1g
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Applications
As active pharmaceutical drugs and intermediates possess different biological activities.

Notes
Moisture Sensitive. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air. Refrigerated (2-4°C).

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.