Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-1-(3-Bromophenyl)ethylamine, ChiPros™ 99%, ee 98+%

Chemical Identifiers

Chemical Name or Material (S)-1-(3-Bromophenyl)ethylamine
CAS 139305-96-7
Molecular Formula C8H10BrN
Formula Weight 200.08
MDL Number MFCD00671638
UN Number UN2735
Synonym (S)-m-Bromo-alpha-methylbenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2699803

  SDS
alfa aesar™
H2699803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2699806

  SDS
alfa aesar™
H2699806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.08
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Optical Rotation −22° (Neat)
Flash Point >110°C (230°F)
CAS 139305-96-7
Synonym (S)-m-Bromo-alpha-methylbenzylamine
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula C8H10BrN
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
UN Number UN2735
Quantity 1g
Assay Percent Notes ee ≥98%
MDL Number MFCD00671638
Assay 99%
Boiling Point 96°C (4mmHg)
Chemical Name or Material (S)-1-(3-Bromophenyl)ethylamine

Applications
(S)-1-(3-Bromophenyl)ethylamine is involved in the Ullmann coupling reaction with N-H containing heteroarenes using copper iodide as a catalyst.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents, carbon monoxide and strong acids.

Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.