Please login to your online account to display your discounted pricing

(S)-(+)-1,2-Propanediol, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-(+)-1, 2-Propanediol
CAS 4254-15-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C3H8O2
Linear Formula CH3CH(OH)CH2OH
Formula Weight 76.1
MDL Number MFCD00004539
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC365300250

  SDS
thermo fisher scientific
AC365300250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC365300050

  SDS
fisher scientific acros
365300050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 76.1
Quantity 25g
Color Clear colorless
Density 102g/mL
Specific Rotation Condition + 16.00 (20.00°C c=neat)
Merck Index 15, 7968
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C3H8O2
Linear Formula CH3CH(OH)CH2OH
MDL Number MFCD00004539
Chemical Name or Material (S)-(+)-1, 2-Propanediol
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
CAS 4254-15-3
Refractive Index 1.4310 to 1.4350 (20°C, 589nm)
Specific Rotation +15° to +18° (20°C, 589nm) (neat)