Please login to your online account to display your discounted pricing

(S)-(+)-1,2-Propanediol, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  365300050

 View more versions of this product

Catalog No. AC365300050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Density 102g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4310 to 1.4350 (20°C, 589nm)
Specific Rotation +15° to +18° (20°C, 589nm) (neat)
Specific Rotation Condition + 16.00 (20.00°C c=neat)
Color Clear colorless
Quantity 5g
Chemical Name or Material (S)-(+)-1, 2-Propanediol
CAS 4254-15-3
CAS Min % 96.0
CAS Max % 100.0
Physical Form Clear colorless liquid
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C3H8O2
Linear Formula CH3CH(OH)CH2OH
Formula Weight 76.10g/mol
MDL Number MFCD00004539
Merck Index 15, 7968

Specifications

Chemical Name or Material (S)-(+)-1, 2-Propanediol
CAS 4254-15-3
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C3H8O2
Linear Formula CH3CH(OH)CH2OH
Formula Weight 76.10g/mol
MDL Number MFCD00004539