Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-1-(2-Naphthyl)ethylamine, ChiPros 99+%, ee 99+%

Chemical Identifiers

Chemical Name or Material (S)-(-)-1-(2-Naphthyl)ethylamine
CAS 3082-62-0
Molecular Formula C12H13N
Formula Weight 171.24
MDL Number MFCD00085366
UN Number UN3259
Beilstein 3195949
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1702403

  SDS
alfa aesar™
L1702403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1702406

  SDS
alfa aesar™
L1702406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.24
Chemical Name or Material (S)-(-)-1-(2-Naphthyl)ethylamine
MDL Number MFCD00085366
CAS 3082-62-0
UN Number UN3259
Beilstein 3195949
Health Hazard 2 GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Assay ≥99%
Assay Percent Notes ee ≥99%
Molecular Formula C12H13N
Health Hazard 1 Danger
Boiling Point 90°C (0.15mmHg)
Melting Point 48° to 52°C
Quantity 1g
Optical Rotation −20° (c=1 in Methanol)

Applications
(S)-(-)-1-(2-Naphthyl)ethylamine is used in asymmetric synthesis. It is also used in the preparation of thiourea compounds using chiral amine, which is used as a bifunctional catalyst for the Strecker and nitro-Michael reactions. Further, it acts as a chiral building block and chiral resolution reagent.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Sensitive to carbon dioxide.

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.