Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-1-(2-Methoxyphenyl)ethylamine, ChiPros™, 99%, ee 98+%

Chemical Identifiers

Chemical Name or Material (S)-1-(2-Methoxyphenyl)ethylamine
CAS 68285-24-5
Molecular Formula C9H13NO
Formula Weight 151.21
MDL Number MFCD00671658
UN Number UN2735
Synonym (S)-o-Methoxy-alpha-methylbenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2694803

  SDS
alfa aesar™
H2694803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2694806

  SDS
alfa aesar™
H2694806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 151.21
Chemical Name or Material (S)-1-(2-Methoxyphenyl)ethylamine
Assay 99%
MDL Number MFCD00671658
Molecular Formula C9H13NO
CAS 68285-24-5
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
UN Number UN2735
Health Hazard 1 Danger
Quantity 1g
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay Percent Notes ee ≥98%
Synonym (S)-o-Methoxy-alpha-methylbenzylamine
Boiling Point 118°C (17mmHg)
Health Hazard 2 GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

Applications
(S)-1-(2-Methoxyphenyl)ethylamine play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks or chiral auxiliaries.

Solubility
Soluble in water(22 g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.