missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-1-[2-(Diphenylphosphino)phenyl]ethylamine, 97+%, Alfa Aesar™

$119.20 - $363.95

Chemical Identifiers

CAS 913196-43-7
Molecular Formula C20H20NP
Molecular Weight (g/mol) 305.361
MDL Number MFCD17013987
InChI Key KPFJIPHGQGHIMM-INIZCTEOSA-N
Synonym s-1-2-diphenylphosphino phenyl ethanamine, s-1-2-diphenylphosphino phenyl ethylamine, s---1-2-diphenylphosphino phenyl ethylamine, s-1-2-diphenylphosphino phenyl ethylamine, s-1-2-diphenylphosphino phenyl ethanamine, 1s-1-2-diphenylphosphanyl phenyl ethanamine, 2-1-aminoethyl triphenylphosphine, s-alpha-methyl-2-diphenylphosphino benzylamine, s-1-2-diphenylphosphanyl phenyl ethan-1-amine, 1s-1-2-diphenylphosphanyl phenyl ethan-1-amine
PubChem CID 57514668
IUPAC Name (1S)-1-(2-diphenylphosphanylphenyl)ethanamine
SMILES CC(C1=CC=CC=C1P(C2=CC=CC=C2)C3=CC=CC=C3)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60122MD View Documents Alfa Aesar
H60122MD
250mg Each for $119.20
 
AAH6012203 View Documents Alfa Aesar
H6012203
1g Each for $363.95
 
Specifications

Chemical Identifiers

913196-43-7
305.361
KPFJIPHGQGHIMM-INIZCTEOSA-N
57514668
CC(C1=CC=CC=C1P(C2=CC=CC=C2)C3=CC=CC=C3)N
C20H20NP
MFCD17013987
s-1-2-diphenylphosphino phenyl ethanamine, s-1-2-diphenylphosphino phenyl ethylamine, s---1-2-diphenylphosphino phenyl ethylamine, s-1-2-diphenylphosphino phenyl ethylamine, s-1-2-diphenylphosphino phenyl ethanamine, 1s-1-2-diphenylphosphanyl phenyl ethanamine, 2-1-aminoethyl triphenylphosphine, s-alpha-methyl-2-diphenylphosphino benzylamine, s-1-2-diphenylphosphanyl phenyl ethan-1-amine, 1s-1-2-diphenylphosphanyl phenyl ethan-1-amine
(1S)-1-(2-diphenylphosphanylphenyl)ethanamine

Specifications

−55° (c=0.5 in Chloroform)
250mg
C20H20NP
s-1-2-diphenylphosphino phenyl ethanamine, s-1-2-diphenylphosphino phenyl ethylamine, s---1-2-diphenylphosphino phenyl ethylamine, s-1-2-diphenylphosphino phenyl ethylamine, s-1-2-diphenylphosphino phenyl ethanamine, 1s-1-2-diphenylphosphanyl phenyl ethanamine, 2-1-aminoethyl triphenylphosphine, s-alpha-methyl-2-diphenylphosphino benzylamine, s-1-2-diphenylphosphanyl phenyl ethan-1-amine, 1s-1-2-diphenylphosphanyl phenyl ethan-1-amine
CC(C1=CC=CC=C1P(C2=CC=CC=C2)C3=CC=CC=C3)N
305.361
305.35
(S)-(-)-1-[2-(Diphenylphosphino)phenyl]ethylamine
76°C to 81°C
913196-43-7
MFCD17013987
KPFJIPHGQGHIMM-INIZCTEOSA-N
(1S)-1-(2-diphenylphosphanylphenyl)ethanamine
57514668
≥97%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger