missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-1-(2,6-Dichloro-3-fluorophenyl)ethanol, 95%

Manufacturer:  Alfa Aesar™ H65374MD

 View more versions of this product

Catalog No. AAH65374MD



Specifications

Specifications

(S)-1-(2,6-Dichloro-3-fluorophenyl)ethanol
250mg
C8H7Cl2FO
1s-1-2,6-dichloro-3-fluoro-phenyl ethanol, 1s-1-2,6-dichloro-3-fluorophenyl ethan-1-ol, 1s-1-2,6-dichloro-3-fluorophenyl ethan-1-ol 30:70 r:s, 1s-1-2,6-dichloro-3-fluorophenyl ethanol, ksc497a2n, pubchem17375, s-1-2',6'-dichloro-3'-fluorophenyl-ethanol, s-1-2,6-dichloro-3-fluorophenyl ethan-1-ol, s-1-2,6-dichloro-3-fluorophenyl ethanol, s-1-2,6-dichloro-3-fluorophenyl-ethanol
CC(C1=C(C=CC(=C1Cl)F)Cl)O
209.041
209.04
95%
White
877397-65-4
MFCD09863793
JAOYKRSASYNDGH-BYPYZUCNSA-N
(1S)-1-(2,6-dichloro-3-fluorophenyl)ethanol
11557536
Solid

Chemical Identifiers

877397-65-4
209.041
JAOYKRSASYNDGH-BYPYZUCNSA-N
11557536
CC(C1=C(C=CC(=C1Cl)F)Cl)O
C8H7Cl2FO
MFCD09863793
1s-1-2,6-dichloro-3-fluoro-phenyl ethanol, 1s-1-2,6-dichloro-3-fluorophenyl ethan-1-ol, 1s-1-2,6-dichloro-3-fluorophenyl ethan-1-ol 30:70 r:s, 1s-1-2,6-dichloro-3-fluorophenyl ethanol, ksc497a2n, pubchem17375, s-1-2',6'-dichloro-3'-fluorophenyl-ethanol, s-1-2,6-dichloro-3-fluorophenyl ethan-1-ol, s-1-2,6-dichloro-3-fluorophenyl ethanol, s-1-2,6-dichloro-3-fluorophenyl-ethanol
(1S)-1-(2,6-dichloro-3-fluorophenyl)ethanol
Safety and Handling

Safety and Handling

P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c

H315-H317-H319-H335

EINECSNumber : 700-699-9

TSCA : No

Recommended Storage : Ambient temperatures

Documents