missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthol, TCI America™

Manufacturer:  TCI America T2360100MG

 View more versions of this product

Catalog No. T2360100MG



Specifications

Specifications

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthol
White-Yellow
100mg
C10H12O
(S)-(+)-alpha-Tetralol
C1CC(C2=CC=CC=C2C1)O
148.205
148.21
≥98.0% (GC)
92°C
41°C
53732-47-1
MFCD00063006
JAAJQSRLGAYGKZ-JTQLQIEISA-N
(1S)-1,2,3,4-tetrahydronaphthalen-1-ol
7058029
Crystal-Powder at 20°C

Chemical Identifiers

53732-47-1
148.205
JAAJQSRLGAYGKZ-JTQLQIEISA-N
7058029
C1CC(C2=CC=CC=C2C1)O
C10H12O
MFCD00063006
(S)-(+)-alpha-Tetralol
(1S)-1,2,3,4-tetrahydronaphthalen-1-ol
Safety and Handling

Safety and Handling

RTECSNumber : QL5075000

TSCA : No

Documents