Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-1-(1-Naphthyl)ethylamine, ChiPros 99+%, ee 99+%

Chemical Identifiers

Chemical Name or Material (S)-(-)-1-(1-Naphthyl)ethylamine
CAS 10420-89-0
Molecular Formula C12H13N
Formula Weight 171.24
MDL Number MFCD00064179
UN Number UN2922
Beilstein 2208024
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1930103

  SDS
alfa aesar™
L1930103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1930106

  SDS
alfa aesar™
L1930106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1930114

  SDS
alfa aesar™
L1930114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.24
Chemical Name or Material (S)-(-)-1-(1-Naphthyl)ethylamine
TSCA TSCA
Assay ≥99%
Assay Percent Notes ee ≥99%
Molecular Formula C12H13N
MDL Number MFCD00064179
CAS 10420-89-0
UN Number UN2922
Beilstein 2208024
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H314-H318-H312
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Odor Amine-like
Recommended Storage 2° to 8°C
Boiling Point 152° to 153°C (11mmHg)
Quantity 1g
Flash Point >110°C (230°F)
Optical Rotation −55° (c=2 in Ethanol)
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H314-H318-H312
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.

2° to 8°C