Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-1-(1-Naphthyl)ethanol, 97%

Chemical Identifiers

Chemical Name or Material (S)-(-)-1-(1-Naphthyl)ethanol
CAS 15914-84-8
Linear Formula C10H7CH(CH3)OH
Formula Weight 172.23
MDL Number MFCD00077831
Beilstein 2045009
Synonym (S)-(-)-α-Methyl-1-naphthalenemethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6010003

  SDS
alfa aesar™
H6010003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 47° to 49°C
Formula Weight 172.23
Physical Form Powder
Color Yellow
Beilstein 2045009
CAS 15914-84-8
MDL Number MFCD00077831
Assay 97%
Linear Formula C10H7CH(CH3)OH
Synonym (S)-(-)-α-Methyl-1-naphthalenemethanol
Quantity 1g
Chemical Name or Material (S)-(-)-1-(1-Naphthyl)ethanol