missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-1,1′-Binaphthyl-2,2′-diyl Bis(trifluoromethanesulfonate), TCI America™

$123.75 - $371.25

Chemical Identifiers

CAS 128544-05-8
Molecular Formula C22H12F6O6S2
Molecular Weight (g/mol) 550.442
MDL Number MFCD00274615
InChI Key OYJLCOSEYYZULE-UHFFFAOYSA-N
Synonym r-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, s-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, r---1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r---1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, s-+-1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r-1,1'-binaphthyl-2,2'-diyl bis trifluoromethane sulphonate, s-+-1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, 1,1'-bi-2-naphthol bis trifluoromethanesulfonate, s-1,1'-bi 2-naphthol bis trifluoromethanesulfonate, r---binol-tf2
PubChem CID 389883
IUPAC Name [1-[2-(trifluoromethylsulfonyloxy)naphthalen-1-yl]naphthalen-2-yl] trifluoromethanesulfonate
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B22311G View Documents TCI America
B22311G
1g Each for $123.75
 
B22315G View Documents TCI America
B22315G
5g Each for $371.25
 
Specifications

Chemical Identifiers

128544-05-8
550.442
OYJLCOSEYYZULE-UHFFFAOYSA-N
389883
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F
C22H12F6O6S2
MFCD00274615
r-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, s-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, r---1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r---1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, s-+-1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r-1,1'-binaphthyl-2,2'-diyl bis trifluoromethane sulphonate, s-+-1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, 1,1'-bi-2-naphthol bis trifluoromethanesulfonate, s-1,1'-bi 2-naphthol bis trifluoromethanesulfonate, r---binol-tf2
[1-[2-(trifluoromethylsulfonyloxy)naphthalen-1-yl]naphthalen-2-yl] trifluoromethanesulfonate

Specifications

White-Yellow
1g
C22H12F6O6S2
3261
OYJLCOSEYYZULE-UHFFFAOYSA-N
[1-[2-(trifluoromethylsulfonyloxy)naphthalen-1-yl]naphthalen-2-yl] trifluoromethanesulfonate
389883
Crystal-Powder at 20°C
(S)-(+)-1,1′-Binaphthyl-2,2′-diyl Bis(trifluoromethanesulfonate)
83°C
128544-05-8
MFCD00274615
r-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, s-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, r---1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r---1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, s-+-1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r-1,1'-binaphthyl-2,2'-diyl bis trifluoromethane sulphonate, s-+-1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, 1,1'-bi-2-naphthol bis trifluoromethanesulfonate, s-1,1'-bi 2-naphthol bis trifluoromethanesulfonate, r---binol-tf2
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F
550.442
550.44
≥93.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No