missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-1,1'-Binaphthol-2,2'-bis(trifluoromethanesulfonate), 97%, ACROS Organics™

Chemical Identifiers

CAS 128544-05-8
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C22 H12 F6 O6 S2
Linear Formula (CF3SO3C10H6-)2
Formula Weight 550.44
MDL Number MFCD00274615
Synonym (S)-1, 1'-Binaphthalene-2, 2'-diyl bis(trifluoromethanesulfonate)
Chemical Name or Material (S)-(+)-1, 1'-Binaphthol-2, 2'-bis(trifluoromethanesulfonate), 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC350440050 SDS Thermo Fisher Scientific
350440050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

128544-05-8
100.0
C22 H12 F6 O6 S2
550.44
(S)-1, 1'-Binaphthalene-2, 2'-diyl bis(trifluoromethanesulfonate)
≥96.0%
96.0
97%
(CF3SO3C10H6-)2
MFCD00274615
(S)-(+)-1, 1'-Binaphthol-2, 2'-bis(trifluoromethanesulfonate), 97%

Specifications

Powder
5g
80° to 85°C
96.0
(CF3SO3C10H6-)2
97%
128544-05-8
+145° to +149° (20°C, 589nm) (c=1, CHCl3)
(S)-(+)-1, 1'-Binaphthol-2, 2'-bis(trifluoromethanesulfonate), 97%
550.44
White to light yellow
100.0
Glass bottle
MFCD00274615
C22 H12 F6 O6 S2
Authentic
(S)-1, 1'-Binaphthalene-2, 2'-diyl bis(trifluoromethanesulfonate)
≥96.0%

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA