Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-1,1'-Bi(2-naphthyl) dimethanesulfonate, 97%

Chemical Identifiers

Chemical Name or Material (S)-(+)-1,1'-Bi(2-naphthyl) dimethanesulfonate
CAS 871231-47-9
Formula Weight 442.5
MDL Number MFCD06798065
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5524903

 
alfa aesar™
H5524903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55249MD

 
alfa aesar™
H55249MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 150° to 154°C
Formula Weight 442.5
Physical Form Solid
Quantity 1g
Chemical Name or Material (S)-(+)-1,1'-Bi(2-naphthyl) dimethanesulfonate
MDL Number MFCD06798065
Assay 97%
CAS 871231-47-9