missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-1,1'-Bi(2-naphthyl) dimethanesulfonate, 97%

Manufacturer:  Alfa Aesar™ H55249MD

 View more versions of this product

Catalog No. AAH55249MD



Specifications

Specifications

(S)-(+)-1,1'-Bi(2-naphthyl) dimethanesulfonate
250mg
MFCD06798065
442.5
97%
150°C to 154°C
871231-47-9
1'-methanesulfonyloxy-2,2'-binaphthalen-1-yl methanesulfonate, r---1,1 inverted exclamation marka-bi-2-naphthyl dimethanesulfonate, r---1,1'-bi-2-naphthyl dimethanesulfonate, s-+-1,1 inverted exclamation marka-bi-2-naphthyl dimethanesulfonate, s-+-1,1'-bi-2-naphthyl dimethanesulfonate, s-+-1,1-bi-2-naphthyl dimethanesul
Solid

Chemical Identifiers

871231-47-9
MFCD06798065
1'-methanesulfonyloxy-2,2'-binaphthalen-1-yl methanesulfonate, r---1,1 inverted exclamation marka-bi-2-naphthyl dimethanesulfonate, r---1,1'-bi-2-naphthyl dimethanesulfonate, s-+-1,1 inverted exclamation marka-bi-2-naphthyl dimethanesulfonate, s-+-1,1'-bi-2-naphthyl dimethanesulfonate, s-+-1,1-bi-2-naphthyl dimethanesul
Safety and Handling

Safety and Handling

P264b-P280i-P305+P351+P338

H319

TSCA : No

Recommended Storage : Ambient temperatures

Documents