missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-1,1'-Bi-2-naphthol, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC224310250

 View more versions of this product

Catalog No. AC224310250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(S)-(-)-1, 1'-Bi-2-naphthol
Glass bottle
-33° to -38° (20°C, 589nm) (c=1, THF)
205°C to 211°C
99%
98.5
99%
HOC10H6C10H6OH
15, 1230
PPTXVXKCQZKFBN-UHFFFAOYSA-N
1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol
11762
Crystals or Crystalline Powder
Authentic
−35.50° (20°C c=1,THF)
White to Gray
25g
18531-99-2
100.0
C20H14O2
16,132
1,1'-bi-2-naphthol
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O
286.33
286.32
≥98.5%

Chemical Identifiers

18531-99-2
286.33
1,1'-bi-2-naphthol
1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol
C20H14O2
PPTXVXKCQZKFBN-UHFFFAOYSA-N
11762
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents