Please login to your online account to display your discounted pricing

(S)-(-)-1,1'-Bi-2-naphthol, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  224310010

 View more versions of this product

Catalog No. AC224310010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystals or Crystalline Powder
Formula Weight 286.32
Assay ≥98.5%
Quantity 1g
Color White to light gray
Fieser 16,132
Merck Index 15, 1230
Melting Point 205° to 211°C
CAS Max % 100.0
Packaging Glass bottle
Assay Percent Range 99%
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C20H14O2
CAS 18531-99-2
Linear Formula HOC10H6C10H6OH
CAS Min % 98.5
Infrared Spectrum Authentic
Specific Rotation -33° to -38° (20°C, 589nm) (c=1, THF)
Name Note 99%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.
Chemical Name or Material (S)-(-)-1, 1'-Bi-2-naphthol
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (S)-(-)-1, 1'-Bi-2-naphthol
Name Note 99%
CAS 18531-99-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C20H14O2
Linear Formula HOC10H6C10H6OH
Formula Weight 286.32
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.

SDS & Safety Documents