Please login to your online account to display your discounted pricing

RWJ 50271, Tocris™

Inhibitor of LFA-1/ICAM mediated cell adhesion

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material RWJ 50271
Assay Percent Range >99%
Molecular Formula C18H17F3N4O2S
Formula Weight 410.41
Synonym N-(3-Hydroxypropyl)-5-methyl-1-[-4-[3-(trifluoromethyl)phenyl)]-2-thiazolyl]-1H-pyrazole-4-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

422710

 
r & d systems
RDS422710
10mg Each for $239.63

422750

 
r & d systems
RDS422750
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 410.41
Quantity 10mg
Assay Percent Range >99%
Synonym N-(3-Hydroxypropyl)-5-methyl-1-[-4-[3-(trifluoromethyl)phenyl)]-2-thiazolyl]-1H-pyrazole-4-carboxamide
Molecular Formula C18H17F3N4O2S
Chemical Name or Material RWJ 50271
CAS 162112-37-0
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature