Please login to your online account to display your discounted pricing

Rutin, 97+%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  132395000

 View more versions of this product

Catalog No. AC132395000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay 97.0 to 102.0% (on dry basis) (Titrimetry other)
Formula Weight 664.58
Quantity 500g
Color White to almost white
Merck Index 15, 8438
EINECS Number 205-814-1
TSCA TSCA
RTECS Number VM2975000
Loss on Drying 5.5 % to 9.0 % (500mg, 130°C, 3 hrs) (vacuum)
Packaging Plastic bottle
Sulfated Ash 0.5 % max.
Health Hazard 1 HYGROSCOPIC
Chemical Name or Material Rutin
Name Note 97.0-102.0%
Assay Percent Range 97+%
Molecular Formula C27H30O16·3H2O
MDL Number MFCD00006830
CAS 153-18-4
Synonym C.I. 75730
Health Hazard 2 LIGHT SENSITIVE
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Rutin
Name Note 97.0-102.0%
CAS 153-18-4
Assay 97.0 to 102.0% (on dry basis) (Titrimetry other)
Assay Percent Range 97+%
Molecular Formula C27H30O16·3H2O
Formula Weight 664.58
MDL Number MFCD00006830
Synonym C.I. 75730
Accessories

HYGROSCOPIC

LIGHT SENSITIVE

SDS & Safety Documents