missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rutin Hydrate, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 132390050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Rutin hydrate | |
207671-50-9 | |
White | |
Authentic | |
Glass Bottle | |
MFCD01319140 | |
15, 8438 | |
Solubility in water: 1g/8l. Other solubilities: soluble in alkaline solutions,soluble in pyridine and formamide,slightly soluble in alcohol and acetone,practically insoluble in chloroform,carbone,disulfide and benzene | |
C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(OC4=CC(O)=CC(O)=C4C3=O)C3=CC(O)=C(O)C=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O | |
610.52 | |
CHEBI:28527 | |
97+% |
97.0-102.0% | |
195.0°C | |
5.5 % to 9.0 % (500mg, 130°C, 3 hrs) (vacuum) | |
97+% | |
C27H30O16 | |
5 g | |
rutin, rutoside, phytomelin, quercetin 3-rutinoside, birutan, eldrin, myrticolorin, venoruton, 3-rutinosyl quercetin, bioflavonoid | |
IKGXIBQEEMLURG-NVPNHPEKSA-N | |
2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one | |
5280805 | |
664.58 | |
Powder |
Chemical Identifiers
207671-50-9 | |
610.52 | |
IKGXIBQEEMLURG-NVPNHPEKSA-N | |
5280805 | |
2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one |
C27H30O16 | |
MFCD01319140 | |
rutin, rutoside, phytomelin, quercetin 3-rutinoside, birutan, eldrin, myrticolorin, venoruton, 3-rutinosyl quercetin, bioflavonoid | |
CHEBI:28527 | |
C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(OC4=CC(O)=CC(O)=C4C3=O)C3=CC(O)=C(O)C=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O |
Safety and Handling
LIGHT SENSITIVE
HYGROSCOPIC
EINECSNumber : 205-814-1
RTECSNumber : VM2975000
TSCA : TSCA
RUO – Research Use Only