missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ruthenium tetroxide, 0.5% solution in water, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 389000100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
Ruthenium tetroxide | |
20427-56-9,7732-18-5 | |
10 mL | |
RuO4 | |
15, 8435 | |
GJFMDWMEOCWXGJ-UHFFFAOYSA-N | |
tetraoxoruthenium | |
119079 | |
0.5 % (typically) | |
1 |
White | |
Crystalline Powder | |
O4Ru | |
MFCD00074857 | |
ruthenium tetroxide, ruthenium tetraoxide, unii-97e960g9rp, ruthenium viii oxide, ruthenium oxide ruo4, sodium perruthenate, ruthenium oxide ruo4 , t-4, ruo4, 51429-86-8 tri-hydrochloride salt | |
O=[Ru](=O)(=O)=O | |
165.07 | |
165.07 | |
Glass Bottle | |
1.0000g/mL |
RUO – Research Use Only