missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ruthenium standard solution, for AAS, 1mg/mL Ru in 5% HCl, Thermo Scientific Chemicals
$56.04 - $56.04
Specifications
Chemical Name or Material | Ruthenium standard solution |
---|---|
Concentration | 990 to 1010μg/mL of Ru(3+) in 10−20% HCl |
Name Note | 1 mg/ml Ru in 5% HCl |
CAS | 7440-18-8,7647-01-0,7732-18-5 |
Packaging | Plastic bottle |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC196251000
|
Thermo Scientific Chemicals
196251000 |
100 mL | Plastic bottle |
Each for $56.04
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopySpecifications
Ruthenium standard solution | |
1 mg/ml Ru in 5% HCl | |
Plastic bottle | |
Ru | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
ruthenium | |
23950 | |
101.07 | |
1.0200g/mL | |
107.0°C | |
Soluble in water |
990 to 1010μg/mL of Ru(3+) in 10−20% HCl | |
7440-18-8,7647-01-0,7732-18-5 | |
Yellow to Brown | |
MFCD00011207 MFCD03458417 | |
[Ru] | |
101.07 | |
CHEBI:30682 | |
AAS | |
-25.0°C | |
1.02 | |
Solution |
RUO – Research Use Only