Please login to your online account to display your discounted pricing

Alfa Aesar™ Ruthenium sponge, -20 mesh, 99.95% (metals basis)

Chemical Identifiers

Chemical Name or Material Ruthenium sponge
CAS 7440-18-8
MDL Number MFCD00011207
UN Number UN3089
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1206204

  SDS
alfa aesar™
1206204
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1206209

  SDS
alfa aesar™
1206209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1206218

  SDS
alfa aesar™
1206218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

TSCA TSCA
Odor Odorless
Assay 99.95%
Physical Form Sponge
Assay Percent Notes (metals basis)
MDL Number MFCD00011207
Health Hazard 2 GHS H Statement
H228
Flammable solid.
Mesh Size −20 Mesh
CAS 7440-18-8
Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
UN Number UN3089
Health Hazard 1 Warning
Quantity 2g
Chemical Name or Material Ruthenium sponge
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

GHS H Statement
H228
Flammable solid.