Please login to your online account to display your discounted pricing

Ruthenium Red tetrahydrate, pure, ACROS Organics™

$217.81 - $217.81

Chemical Identifiers

Chemical Name or Material Ruthenium red tetrahydrate
Name Note Pure
CAS 12790-48-6
Purity Grade Extra Pure
Assay 34.6 to 36.1% (ruthenium)
Molecular Formula Cl6H42N14O2Ru3·4H2O
Linear Formula {(NH3)5Ruoru(NH3)4oru(NH3)5}Cl6·4H2O
Formula Weight 858.35
MDL Number MFCD00167176
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC191730050

  SDS
thermo fisher scientific
AC191730050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC191730010

  SDS
fisher scientific acros
191730010
Glass bottle 1g Each for $217.81
Description & Specifications

Specifications

Formula Weight 858.35
Purity Grade Extra Pure
Physical Form Crystalline Powder
Quantity 5g
Color Dark red-purple
Assay 34.6 to 36.1% (ruthenium)
Merck Index 10, 8161
Lambda Max 530 to 536nm
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Molar Absorption Coefficient 8000L/(mole.cm) (lambda min., 0.02g/L H2O)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Ruthenium red tetrahydrate
Linear Formula {(NH3)5Ruoru(NH3)4oru(NH3)5}Cl6·4H2O
Molecular Formula Cl6H42N14O2Ru3·4H2O
CAS 12790-48-6
Name Note Pure
MDL Number MFCD00167176
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.