Learn More
Ruthenium, 10% on activated carbon powder, reduced, Thermo Scientific Chemicals
$487.37 - $1888.47
Chemical Identifiers
CAS | 7440-18-8 |
---|---|
Molecular Formula | Ru |
Molecular Weight (g/mol) | 101.07 |
MDL Number | MFCD00011207 MFCD03458417 |
InChI Key | KJTLSVCANCCWHF-UHFFFAOYSA-N |
Synonym | on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon |
PubChem CID | 23950 |
ChEBI | CHEBI:30682 |
IUPAC Name | ruthenium |
SMILES | [Ru] |
Description
Ruthenium, 10% on activated carbon powder is used in hydrogenation of aromatic ring systems, particularly the hindered systems. It finds wide use for the reduction of aliphatic carbonyl compounds and carboxylic acids to alcohols, hydrogenation of anilines to cyclohexylamines, synthesis of ammonia, depolymerization of carbohydrates, Fischer-Tropsch synthesis, and hydrogenation of glucose to produce sorbitol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-18-8 | |
101.07 | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
23950 | |
ruthenium |
Ru | |
MFCD00011207 MFCD03458417 | |
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon | |
CHEBI:30682 | |
[Ru] |
Specifications
Black | |
Black powder | |
MFCD00011207 MFCD03458417 | |
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
ruthenium | |
23950 | |
Odorless | |
Powder |
7440-18-8 | |
Ru | |
25 g | |
Insoluble in water. | |
[Ru] | |
101.07 | |
CHEBI:30682 | |
Air Sensitive | |
Ruthenium |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
P210-P235+P410-P240-P241-P280-P370+P378q
H228-H252
DOTInformation : DOT Class: 4.2, Packaging Group: III
EINECSNumber : 231-127-1
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only