missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ruthenium, 10% on activated carbon powder, reduced, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01175722

 View more versions of this product

Catalog No. AA1175722



Description

Description

Ruthenium, 10% on activated carbon powder is used in hydrogenation of aromatic ring systems, particularly the hindered systems. It finds wide use for the reduction of aliphatic carbonyl compounds and carboxylic acids to alcohols, hydrogenation of anilines to cyclohexylamines, synthesis of ammonia, depolymerization of carbohydrates, Fischer-Tropsch synthesis, and hydrogenation of glucose to produce sorbitol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Black
7440-18-8
Ru
100 g
Insoluble in water.
[Ru]
101.07
CHEBI:30682
Air Sensitive
Ruthenium
Black powder
MFCD00011207 MFCD03458417
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
KJTLSVCANCCWHF-UHFFFAOYSA-N
ruthenium
23950
Odorless
Powder

Chemical Identifiers

7440-18-8
101.07
KJTLSVCANCCWHF-UHFFFAOYSA-N
23950
ruthenium
Ru
MFCD00011207 MFCD03458417
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
CHEBI:30682
[Ru]
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.

P210-P235+P410-P240-P241-P280-P370+P378q

H228-H252

DOTInformation : DOT Class: 4.2, Packaging Group: III

EINECSNumber : 231-127-1

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only