missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ruthenium powder, 99.>99% (metals basis excluding Ca), Alfa Aesar™, Premion™

Manufacturer:  Alfa Aesar 4487903

 View more versions of this product

Catalog No. AA4487903



Specifications

Specifications

Ruthenium
7440-18-8
>99.99% (Metals basis excluding Ca)
MFCD00011207
KJTLSVCANCCWHF-UHFFFAOYSA-N
[Ru]
101.07
CHEBI:30682
Gray
1g
gray powder
Ru
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
Odorless
ruthenium
23950
Powder

Chemical Identifiers

7440-18-8
101.07
KJTLSVCANCCWHF-UHFFFAOYSA-N
23950
ruthenium
Ru
MFCD00011207
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
CHEBI:30682
[Ru]
Safety and Handling

Safety and Handling

H228: Flammable solid.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-127-1

SDS Documents