missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ruthenium powder, -22 mesh, Premion™, 99.98% (metals basis excluding Ca), Ca 100ppm max

Manufacturer:  Alfa Aesar™ 1066706

 View more versions of this product

Catalog No. AA1066706



Specifications

Specifications

Ruthenium powder
(metals basis excluding Ca), Ca 100ppm max.
MFCD00011207
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
[Ru]
101.07
CHEBI:30682
99.98%
−22 Mesh
5g
7440-18-8
Ru
UN3089
KJTLSVCANCCWHF-UHFFFAOYSA-N
ruthenium
23950
Powder
Premion™
Odorless

Chemical Identifiers

7440-18-8
101.07
KJTLSVCANCCWHF-UHFFFAOYSA-N
23950
ruthenium
Ru
MFCD00011207
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
CHEBI:30682
[Ru]
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

TSCA : TSCA

Documents