Please login to your online account to display your discounted pricing

Alfa Aesar™ Ruthenium pellets, 3-6mm, 99.95% (metals basis)

Chemical Identifiers

Chemical Name or Material Ruthenium pellets
CAS 7440-18-8
MDL Number MFCD00011207
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA5523606

 
alfa aesar™
5523606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Pellet
Chemical Name or Material Ruthenium pellets
TSCA TSCA
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD00011207
CAS 7440-18-8
Odor Odorless
Particle Size 3 to 6mm
Color Gray
Weight ≈1.5 to 2.5g/pellet
Quantity 5g