Please login to your online account to display your discounted pricing

Ruthenium(IV) oxide hydrate, Ru 54% min., Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Ruthenium(IV) oxide hydrate
CAS 32740-79-7
Physical Form Powder
Assay Percent Range Ru 54% min.
Molecular Formula O2Ru·xH2O
Formula Weight 133.07(anhydrous)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1180303

  SDS
alfa aesar
1180303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1180306

  SDS
alfa aesar
1180306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1180314

  SDS
alfa aesar
1180314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Ruthenium(IV) oxide hydrate
CAS 32740-79-7
Health Hazard 1 H271: May cause fire or explosion; strong oxidizer.
DOT Information DOT Class: 5.1, Packaging Group: II
EINECS Number 234-840-6
Physical Form Powder
Assay Percent Range Ru 54% min.
Molecular Formula O2Ru·xH2O
Appearance Black powder
Formula Weight 133.07(anhydrous)
Odor Odorless
Color Black
Quantity 1g
Safety and Handling

H271: May cause fire or explosion; strong oxidizer.