missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ruthenium(IV) oxide hydrate, > 54% Ru, Thermo Scientific Chemicals
$561.05 - $2453.92
Chemical Identifiers
CAS | 32740-79-7 |
---|---|
Molecular Formula | O2Ru |
Molecular Weight (g/mol) | 133.07 |
MDL Number | MFCD00149846 |
InChI Key | WOCIAKWEIIZHES-UHFFFAOYSA-N |
IUPAC Name | dioxoruthenium |
SMILES | O=[Ru]=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC197430050
|
Thermo Scientific Chemicals
197430050 |
5 g | Glass bottle |
Each for $561.05
|
|
||||
AC197430250
|
Thermo Scientific Chemicals
197430250 |
25 g | Glass bottle |
Each for $2,453.92
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
32740-79-7 | |
133.07 | |
WOCIAKWEIIZHES-UHFFFAOYSA-N | |
O=[Ru]=O |
O2Ru | |
MFCD00149846 | |
dioxoruthenium |
Specifications
Black | |
Fine Powder | |
O2Ru | |
MFCD00149846 | |
Solubility in water: insoluble | |
O=[Ru]=O | |
133.07 | |
>54% (Ru) | |
Ruthenium(IV) oxide hydrate, >54% Ru |
32740-79-7 | |
5 g | |
RuO2·xH2O | |
01,984; 14,272; 04,108; 05,161 | |
WOCIAKWEIIZHES-UHFFFAOYSA-N | |
dioxoruthenium | |
133.07 | |
Glass bottle |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO – Research Use Only