Please login to your online account to display your discounted pricing

Ruthenium(IV) oxide, Electronic Grade, 99.95% (Metals basis), Alfa Aesar™, Premion™

For Semiconductor Industry Use For the Manufacture of Resistor Pastes

Chemical Identifiers

Chemical Name or Material Ruthenium(IV) oxide
CAS 12036-10-1
Purity Grade Electronic Grade
Physical Form Powder
Assay Percent Range 99.95% (metals basis)
Molecular Formula RuO2
Formula Weight 133.07
MDL Number MFCD00011210
Synonym Ruthenium dioxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4033602

  SDS
alfa aesar
4033602
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4033604

  SDS
alfa aesar
4033604
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4033609

  SDS
alfa aesar
4033609
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Electronic Grade
Physical Form Powder
Formula Weight 133.07
Quantity 0.5g
Color Black
DOT Information DOT Class: 5.1, Packaging Group: II
Chemical Name or Material Ruthenium(IV) oxide
EINECS Number 234-840-6
Odor Odorless
Assay Percent Range 99.95% (metals basis)
Molecular Formula RuO2
Appearance Black powder
MDL Number MFCD00011210
CAS 12036-10-1
Synonym Ruthenium dioxide
Health Hazard 1 H271-H303: May cause fire or explosion; strong oxidizer. May be harmful if swallowed.
Safety and Handling

H271-H303: May cause fire or explosion; strong oxidizer. May be harmful if swallowed.