missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ruthenium(IV) oxide, anhydrous, 99.95% (metals basis), Ru 75.2% min., Alfa Aesar™ Premion™

Manufacturer:  Alfa Aesar 1180404

 View more versions of this product

Catalog No. AA1180404



Specifications

Specifications

Ruthenium(IV) oxide, anhydrous
12036-10-1
O2Ru
Black powder
WOCIAKWEIIZHES-UHFFFAOYSA-N
dioxoruthenium
82848
Powder
Odorless
2g
Ru 75.2% min.
99.95% (metals basis)
MFCD00011210
ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, ruthenium iv oxide, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous
O=[Ru]=O
133.068
133.07
6.97g/cm3 at 20°C
Black

Chemical Identifiers

12036-10-1
133.068
WOCIAKWEIIZHES-UHFFFAOYSA-N
82848
O=[Ru]=O
O2Ru
MFCD00011210
ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, ruthenium iv oxide, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous
dioxoruthenium
Safety and Handling

Safety and Handling

H271-H303: May cause fire or explosion; strong oxidizer. May be harmful if swallowed.

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 234-840-6

Documents