Learn More
Ruthenium(IV) oxide, anhydrous, 99.9%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1081606
Description
The most often used oxygen catalyst is ruthenium(IV) oxide. It is applied as an electrochemical super capacitor material, and it has high capacity to store charge. RuO2 is used as catalyst in the Deacon process which produces chlorine through the oxidation of hydrogen chloride. Owing to the high charge transfer capability, it is used as active material in super capacitors. The dispersion property of RuO2 is used to increase the capacitance of nanocomposite electrode materials for supercapacitors. RuO2 is extensively used for the coating of titanium anodes, and as a catalyst and doping agent, for use in the removal of hydrogen sulfide from oil refineries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe most often used oxygen catalyst is ruthenium(IV) oxide. It is applied as an electrochemical super capacitor material, and it has high capacity to store charge. RuO2 is used as catalyst in the Deacon process which produces chlorine through the oxidation of hydrogen chloride. Owing to the high charge transfer capability, it is used as active material in super capacitors. The dispersion property of RuO2 is used to increase the capacitance of nanocomposite electrode materials for supercapacitors. RuO2 is extensively used for the coating of titanium anodes, and as a catalyst and doping agent, for use in the removal of hydrogen sulfide from oil refineries.
Solubility
Insoluble in water, and acids. Soluble in fused alkali.
Notes
Incompatible with oxidizers, acids, aqua regia, organic solvents.
Specifications
Ruthenium (IV) oxide | |
Anhydrous | |
5 g | |
O2Ru | |
UN1479 | |
Insoluble in water,and acids. Soluble in fused alkali. | |
O=[Ru]=O | |
133.07 | |
133.07 | |
Odorless | |
6.97 |
1200°C (sublimation) | |
12036-10-1 | |
(metals basis) | |
MFCD00011210 | |
ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous | |
WOCIAKWEIIZHES-UHFFFAOYSA-N | |
dioxoruthenium | |
82848 | |
99.9% | |
Hygroscopic |
Chemical Identifiers
12036-10-1 | |
133.07 | |
WOCIAKWEIIZHES-UHFFFAOYSA-N | |
82848 | |
O=[Ru]=O |
O2Ru | |
MFCD00011210 | |
ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous | |
dioxoruthenium |
Safety and Handling
GHS H Statement
H271
May cause fire or explosion
strong oxidizer.
P264b-P280i-P305+P351+P338
H319
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber : 234-840-6
RTECSNumber : VM2654000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only