Please login to your online account to display your discounted pricing

Ruthenium(IV) oxide, 99.5+%, anhydrous, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  218340010

 View more versions of this product

Catalog No. AC218340010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.50%
Physical Form Fine Powder
Formula Weight 133.07
Quantity 1g
Color Black
TSCA TSCA
RTECS Number VM2654000
EINECS Number 234-840-6
Density 6.97g/mL
Fieser 12,428; 14,272; 15,281
MDL Number MFCD00011210
Name Note 99.5+%
Linear Formula RuO2
Assay Percent Range 99.5+%
Packaging Glass bottle
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula O2Ru
Health Hazard 2 GHS P Statement
Causes serious eye irritation.
Chemical Name or Material Ruthenium(IV) oxide
CAS 12036-10-1
Health Hazard 1 HYGROSCOPIC

Specifications

Chemical Name or Material Ruthenium(IV) oxide
Name Note 99.5+%
CAS 12036-10-1
Assay ≥99.50%
Assay Percent Range 99.5+%
Molecular Formula O2Ru
Linear Formula RuO2
Formula Weight 133.07
MDL Number MFCD00011210
Accessories

HYGROSCOPIC

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes serious eye irritation.

SDS & Safety Documents