missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v

Manufacturer:  Alfa Aesar™ 12630AD

 View more versions of this product

Catalog No. AA12630AD



Specifications

Specifications

Ruthenium(III) nitrosylnitrate solution
Ru(NO)(NO3)3
UN3093
1.07g/mL
Liquid
Red to Brown
34513-98-9
MFCD00016313
Ru 1.5% w/v
317.09
Acidic
50mL

Chemical Identifiers

34513-98-9
MFCD00016313
Safety and Handling

Safety and Handling

GHS H Statement
H272-H314
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, OXIDIZING, N.O.S.

TSCA : TSCA

Documents