Please login to your online account to display your discounted pricing

Ruthenium(III) nitrosyl nitrate, in diluted nitric acid, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC365045000

 View more versions of this product

Catalog No. AC365045000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage. May be corrosive to metals.
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Physical Form Solution
Formula Weight 317.09
Quantity 500mL
Color Clear orange to dark brown
Assay 1.5% Ru (typically)
Density 1.0300g/mL
Packaging Glass bottle
CAS 34513-98-9
Molecular Formula N4O10Ru
Name Note In diluted nitric acid
Chemical Name or Material Ruthenium(III) nitrosyl nitrate
MDL Number MFCD00016313
Health Hazard 1 Danger

Specifications

Chemical Name or Material Ruthenium(III) nitrosyl nitrate
Name Note In diluted nitric acid
CAS 34513-98-9
Assay 1.5% Ru (typically)
Molecular Formula N4O10Ru
Formula Weight 317.09
MDL Number MFCD00016313
Accessories

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
Causes severe skin burns and eye damage. May be corrosive to metals.

SDS & Safety Documents