Please login to your online account to display your discounted pricing

Ruthenium(III) nitrosyl nitrate, in diluted nitric acid, ACROS Organics™


Quantity 25mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC365040250

 View more versions of this product

Catalog No. AC365040250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 317.09
Quantity 25mL
Color Clear orange to dark brown
Assay 1.5% Ru (typically)
Density 1.0300g/mL
Packaging Glass bottle
CAS 34513-98-9
Chemical Name or Material Ruthenium(III) nitrosyl nitrate
Name Note In diluted nitric acid
Molecular Formula N4O10Ru
MDL Number MFCD00016313
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. May be corrosive to metals.

Specifications

Chemical Name or Material Ruthenium(III) nitrosyl nitrate
Name Note In diluted nitric acid
CAS 34513-98-9
Assay 1.5% Ru (typically)
Molecular Formula N4O10Ru
Formula Weight 317.09
MDL Number MFCD00016313
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

GHS P Statement Causes severe skin burns and eye damage. May be corrosive to metals.

SDS & Safety Documents