Please login to your online account to display your discounted pricing

Ruthenium(III) Chloride Hydrate, EMD Millipore


Manufacturer: emd millipore corporation  1192470005

 View more versions of this product

Catalog No. M1192470005

  • / Each
 

Description & Specifications

Chemical Identifiers

DOT Information DOT Class: 8, Packaging Group: II
Chemical Name or Material Ruthenium(III) chloride hydrate
EINECS Number 233-167-5
Odor Of Hydrochloric acid
Packaging Glass bottle
Physical Form Powder
Molecular Formula RuCl3·xH2O
Appearance Brown powder
CAS 14898-67-0
UN Number UN 3260
Health Hazard 1 H302: Harmful if swallowed.
H314: Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material Ruthenium(III) chloride hydrate
CAS 14898-67-0
Molecular Formula RuCl3·xH2O
UN Number UN 3260
Accessories

H302: Harmful if swallowed.
H314: Causes severe skin burns and eye damage.