Please login to your online account to display your discounted pricing

Ruthenium(III) Chloride Hydrate, EMD Millipore

$555.91 - $2,225.51

Chemical Identifiers

Chemical Name or Material Ruthenium(III) chloride hydrate
CAS 14898-67-0
Molecular Formula RuCl3·xH2O
UN Number UN 3260
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

M1192470005

 
emd millipore corporation
1192470005
Each for $555.91

M1192470025

 
emd millipore corporation
1192470025
Each for $2,225.51
Description & Specifications

Specifications

DOT Information DOT Class: 8, Packaging Group: II
EINECS Number 233-167-5
Appearance Brown powder
CAS 14898-67-0
UN Number UN 3260
Physical Form Powder
Odor Of Hydrochloric acid
Chemical Name or Material Ruthenium(III) chloride hydrate
Packaging Glass bottle
Molecular Formula RuCl3·xH2O
Health Hazard 1 H302: Harmful if swallowed.
H314: Causes severe skin burns and eye damage.
Safety and Handling

H302: Harmful if swallowed.
H314: Causes severe skin burns and eye damage.