Please login to your online account to display your discounted pricing

Ruthenium(III) Chloride Hydrate, EMD Millipore


Manufacturer: emd millipore corporation  1192470025

 View more versions of this product

Catalog No. M1192470025

  • / Each
 

Description & Specifications

Chemical Identifiers

DOT Information DOT Class: 8, Packaging Group: II
Health Hazard 1 H302: Harmful if swallowed.
H314: Causes severe skin burns and eye damage.
CAS 14898-67-0
Molecular Formula RuCl3·xH2O
Odor Of Hydrochloric acid
Chemical Name or Material Ruthenium(III) chloride hydrate
Appearance Brown powder
UN Number UN 3260
Packaging Glass bottle
EINECS Number 233-167-5
Physical Form Powder

Specifications

Chemical Name or Material Ruthenium(III) chloride hydrate
CAS 14898-67-0
Molecular Formula RuCl3·xH2O
UN Number UN 3260
Accessories

H302: Harmful if swallowed.
H314: Causes severe skin burns and eye damage.