missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ruthenium(III) chloride hydrate, 99.9% (PGM basis), Ru 38% min, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1104309

 View more versions of this product

Catalog No. AA1104309



Specifications

Specifications

Ruthenium(III) chloride hydrate
14898-67-0
Cl3Ru·xH2O
Black crystalline or powder
Crystalline
10g
PGM basis, Ru 38% min.
99.9%
MFCD00149844
207.43 (Anhydrous)
Pungent

Chemical Identifiers

14898-67-0
Cl3Ru·xH2O
MFCD00149844
Safety and Handling

Safety and Handling

H314-H318-H290-H302-H412: Causes severe skin burns and eye damage. Causes serious eye damage. May be corrosive to metals. Harmful if swallowed. Harmful to aquatic life with long lasting effects.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 233-167-5

Documents