missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ruthenium(III) chloride hydrate, 99.9% (PGM basis), Ru 38% min, Alfa Aesar™

$118.28 - $1,774.35

Chemical Identifiers

CAS 14898-67-0
Molecular Formula Cl3Ru·xH2O
MDL Number MFCD00149844
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1104304
View Documents
Thermo Fisher Scientific
1104304
2g
Each for $118.28
 
AA1104309
View Documents
Thermo Fisher Scientific
1104309
10g
Each for $474.12
 
AA1104318
View Documents
Thermo Fisher Scientific
1104318
50g
Each for $1,774.35
 
Description

Description

Specifications

Chemical Identifiers

14898-67-0
Cl3Ru·xH2O
MFCD00149844

Specifications

14898-67-0
Cl3Ru·xH2O
Black crystalline or powder
207.43 (Anhydrous)
Pungent
100°dec
Ruthenium(III) chloride hydrate, PGM basis, Ru 38% min.
99.9%
MFCD00149844
Very soluble in water. Soluble in alcohol,acetone.
Crystalline Soluble
Hygroscopic
2g
Safety and Handling

Safety and Handling

P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c

H290-H302-H314-H335

DOTInformation : DOT Class: 8; Packaging Group: II

EINECSNumber : 233-167-5

RTECSNumber : VM2650000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS