missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ruthenium(III) chloride hydrate, 99.9% (PGM basis), Ru 38% min, Alfa Aesar™
Manufacturer: Thermo Fisher Scientific 1104318
Specifications
Ruthenium(III) chloride hydrate | |
14898-67-0 | |
Cl3Ru·xH2O | |
Black crystalline or powder | |
207.43 (Anhydrous) | |
Pungent | |
100°dec |
PGM basis, Ru 38% min. | |
99.9% | |
MFCD00149844 | |
Very soluble in water. Soluble in alcohol,acetone. | |
Crystalline Soluble | |
Hygroscopic | |
50g |
Chemical Identifiers
14898-67-0 | |
Cl3Ru·xH2O | |
MFCD00149844 |
Safety and Handling
P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H302-H314-H335
DOTInformation : DOT Class: 8; Packaging Group: II
EINECSNumber : 233-167-5
RTECSNumber : VM2650000
TSCA : Yes
Recommended Storage : Ambient temperatures