missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ruthenium(III) chloride hydrate, 99.9% (PGM basis), Ru 38% min, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1104304

 View more versions of this product

Catalog No. AA1104304



Specifications

Specifications

Ruthenium(III) chloride hydrate
14898-67-0
Cl3Ru·xH2O
Black crystalline or powder
207.43 (Anhydrous)
Pungent
100°dec
PGM basis, Ru 38% min.
99.9%
MFCD00149844
Very soluble in water. Soluble in alcohol,acetone.
Crystalline Soluble
Hygroscopic
2g

Chemical Identifiers

14898-67-0
Cl3Ru·xH2O
MFCD00149844
Safety and Handling

Safety and Handling

P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c

H290-H302-H314-H335

DOTInformation : DOT Class: 8; Packaging Group: II

EINECSNumber : 233-167-5

RTECSNumber : VM2650000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents