missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ruthenium(III) chloride, anhydrous, Ru 47.7% min

Manufacturer:  Alfa Aesar™ 4718204*

 View more versions of this product

Catalog No. AA4718204



Specifications

Specifications

Ruthenium(III) chloride
10049-08-8
MFCD00011208
14,8302
Powder
3.110g/mL
Hygroscopic
>500°C (decomposition)
Anhydrous
RuCl3
UN3260
207.43
Ru 47.7% min
Pungent
Black
2g

Chemical Identifiers

10049-08-8
MFCD00011208
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H290-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

RTECSNumber : VM2650000

TSCA : TSCA

SDS Documents