missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ruthenium(III) bromide hydrate, Ru 25% min

Oxidation

Manufacturer:  Alfa Aesar™ 1262803

 View more versions of this product

Catalog No. AA1262803



Specifications

Specifications

Ruthenium(III) bromide hydrate
MFCD00016314
Soluble in alcohol
Crystalline
Characteristic
1g
314075-07-5
UN3260
340.78 (Anhydrous)
Ru 25% min.
Hygroscopic

Chemical Identifiers

314075-07-5
MFCD00016314
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 237-829-4

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents