Learn More
Ruthenium(III) bromide hydrate, Ru 25% min, Thermo Scientific Chemicals
Oxidation
Supplier: Thermo Scientific Chemicals 01262806
Description
Ruthenium(III) bromide hydrate is used as a catalyst in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ruthenium(III) bromide hydrate | |
Crystalline | |
MFCD00016314 | |
Soluble in alcohol | |
Ru 25% min. | |
Hygroscopic |
314075-07-5 | |
5 g | |
UN3260 | |
340.78 (Anhydrous) | |
Characteristic |
Chemical Identifiers
314075-07-5 | |
MFCD00016314 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 237-829-4
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only